siRNA Details
| Gene Symbol: Cabyr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143893.1 | NP_001137365.1 | 5 | 1417 | ||
| XM_006254426.2 | XP_006254488.1 | 5 | 2706 | |||
| XM_008771955.1 | XP_008770177.1 | 5 | 2706 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cabyr| Gene Symbol: Cabyr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143893.1 | NP_001137365.1 | 5 | 1417 | ||
| XM_006254426.2 | XP_006254488.1 | 5 | 2706 | |||
| XM_008771955.1 | XP_008770177.1 | 5 | 2706 | |||