siRNA Details
| Gene Symbol: LOC686209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750016.1 | XP_003750064.1 | 745 | |||
| Gene Symbol: LOC100912529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750010.1 | XP_003750058.1 | 1 | 745 | ||

LOC686209
Genomic Map