siRNA Details
| Gene Symbol: LOC688828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109516.1 | NP_001102986.1 | 3 | 683 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC688828| Gene Symbol: LOC688828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109516.1 | NP_001102986.1 | 3 | 683 | ||