siRNA Details
| Gene Symbol: Naa16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751517.3 | XP_003751565.1 | 2 | 256 | ||
| XM_008770925.1 | XP_008769147.1 | 2 | 256 | |||
| XM_008770926.1 | XP_008769148.1 | 2 | 256 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naa16| Gene Symbol: Naa16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751517.3 | XP_003751565.1 | 2 | 256 | ||
| XM_008770925.1 | XP_008769147.1 | 2 | 256 | |||
| XM_008770926.1 | XP_008769148.1 | 2 | 256 | |||