siRNA Details
| Gene Symbol: Fcrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232673.2 | XP_006232735.1 | 3 | 1228 | ||
| XM_006232674.2 | XP_006232736.1 | 2 | 1207 | |||
| GenBank mRNA | FQ232638.1 | - | 3 | 159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcrl1| Gene Symbol: Fcrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232673.2 | XP_006232735.1 | 3 | 1228 | ||
| XM_006232674.2 | XP_006232736.1 | 2 | 1207 | |||
| GenBank mRNA | FQ232638.1 | - | 3 | 159 | ||