siRNA Details
| Gene Symbol: Tpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134994.1 | NP_001128466.1 | 4, 5 | 348 | ||
| XM_008762921.1 | XP_008761143.1 | 1, 2 | 1894 | |||
| GenBank mRNA | BC168695.1 | - | 4, 5 | 237 | 7938123 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpk1| Gene Symbol: Tpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134994.1 | NP_001128466.1 | 4, 5 | 348 | ||
| XM_008762921.1 | XP_008761143.1 | 1, 2 | 1894 | |||
| GenBank mRNA | BC168695.1 | - | 4, 5 | 237 | 7938123 | |