siRNA Details
| Gene Symbol: LOC100912041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FQ229335.1 | - | 6, 7 | 283 | ||
| FQ229630.1 | - | 6, 7 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100912041| Gene Symbol: LOC100912041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FQ229335.1 | - | 6, 7 | 283 | ||
| FQ229630.1 | - | 6, 7 | 283 | |||