siRNA Details
| Gene Symbol: Rnpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100810.1 | NP_001094280.1 | 1 | 222 | ||
| XM_006233211.2 | XP_006233273.1 | 1 | 600 | |||
| XM_008761424.1 | XP_008759646.1 | 1 | 600 | |||
| XM_008761425.1 | XP_008759647.1 | 1 | 601 | |||
| GenBank mRNA | BC166571.1 | - | 1 | 222 | 7320328 | |

Rnpc3
Genomic Map