siRNA Details
| Gene Symbol: KIAA0586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244189.1 | NP_001231118.1 | 28 | 4297 | ||
| NM_001244190.1 | NP_001231119.1 | 27 | 4140 | |||
| NM_001244191.1 | NP_001231120.1 | 26 | 3948 | |||
| NM_001244192.1 | NP_001231121.1 | 27 | 4071 | |||
| NM_001244193.1 | NP_001231122.1 | 23 | 3584 | |||
| NM_014749.3 | NP_055564.3 | 25 | 3939 | |||
| GenBank mRNA | AB011158.1 | - | 27 | 4123 | ||
| AK302579.1 | - | 23 | 3586 | |||
| AK302635.1 | - | 26 | 3907 | |||
| AK302836.1 | - | 28 | 4297 | |||
| AK310356.1 | - | 23 | 3600 | |||
| AY359881.1 | - | 27 | 4020 | |||
| BC000900.2 | - | 3 | 292 | |||
| BC066647.1 | - | 25 | 3894 | 6166031 | ||

KIAA0586
Genomic Map