siRNA Details
| Gene Symbol: Lmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749863.3 | XP_003749911.1 | 2 | 650 | ||
| XM_006237585.2 | XP_006237647.1 | 2 | 634 | |||
| XM_006237588.2 | XP_006237650.1 | 3 | 642 | |||
| XM_006237590.2 | XP_006237652.1 | 2 | 248 | |||
| XM_006237591.2 | XP_006237653.1 | 2 | 428 | |||
| XM_008763453.1 | XP_008761675.1 | 2 | 578 | |||
| GenBank mRNA | FQ213730.1 | - | 1 | 393 | ||

Lmo3
Genomic Map