siRNA Details
| Gene Symbol: KIAA0586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244189.1 | NP_001231118.1 | 23 | 3495 | ||
| NM_001244190.1 | NP_001231119.1 | 22 | 3338 | |||
| NM_001244191.1 | NP_001231120.1 | 21 | 3146 | |||
| NM_001244192.1 | NP_001231121.1 | 22 | 3269 | |||
| NM_001244193.1 | NP_001231122.1 | 18 | 2782 | |||
| NM_014749.3 | NP_055564.3 | 20 | 3137 | |||
| GenBank mRNA | AB011158.1 | - | 22 | 3321 | ||
| AK302579.1 | - | 18 | 2784 | |||
| AK302635.1 | - | 21 | 3105 | |||
| AK302718.1 | - | 18 | 2787 | |||
| AK302836.1 | - | 23 | 3495 | |||
| AK310356.1 | - | 18 | 2798 | |||
| AY359881.1 | - | 22 | 3218 | |||
| BC066647.1 | - | 20 | 3092 | 6166031 | ||

KIAA0586
Genomic Map