siRNA Details
| Gene Symbol: LOC686288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101012.1 | NP_001094482.1 | 4 | 840 | ||
| GenBank mRNA | BC128716.1 | - | 4 | 840 | 7128779 | |
| Gene Symbol: Olr1668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000267.1 | NP_001000267.1 | 1 | 358 | ||

LOC686288
Genomic Map