siRNA Details
| Gene Symbol: Tmem223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191104.1 | NP_001178033.1 | 2 | 567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem223| Gene Symbol: Tmem223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191104.1 | NP_001178033.1 | 2 | 567 | ||