siRNA Details
| Gene Symbol: Ephb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751156.3 | XP_003751204.1 | 7 | 1928 | ||
| XM_003751157.3 | XP_003751205.1 | 7 | 1955 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ephb4| Gene Symbol: Ephb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751156.3 | XP_003751204.1 | 7 | 1928 | ||
| XM_003751157.3 | XP_003751205.1 | 7 | 1955 | |||