siRNA Details
| Gene Symbol: Trip6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127570.1 | NP_001121042.1 | 6 | 1017 | ||
| GenBank mRNA | BC161996.1 | - | 6 | 1017 | 9034684 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trip6| Gene Symbol: Trip6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127570.1 | NP_001121042.1 | 6 | 1017 | ||
| GenBank mRNA | BC161996.1 | - | 6 | 1017 | 9034684 | |