siRNA Details
| Gene Symbol: LOC690344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168661.1 | NP_001162132.1 | 2 | 599 | ||
| GenBank mRNA | FM119083.1 | - | 2 | 599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690344| Gene Symbol: LOC690344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168661.1 | NP_001162132.1 | 2 | 599 | ||
| GenBank mRNA | FM119083.1 | - | 2 | 599 | ||