siRNA Details
| Gene Symbol: Pramef17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001076970.3 | XP_001076970.1 | 2 | 730 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pramef17| Gene Symbol: Pramef17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001076970.3 | XP_001076970.1 | 2 | 730 | ||