siRNA Details
| Gene Symbol: Wdr74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109569.1 | NP_001103039.1 | 4 | 374 | ||
| XM_006230999.2 | XP_006231061.1 | 4 | 381 | |||
| XM_006231000.2 | XP_006231062.1 | 4 | 381 | |||
| GenBank mRNA | BC158834.1 | - | 4 | 343 | 7383724 | |

Wdr74
Genomic Map