siRNA Details
| Gene Symbol: Lsmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001071401.3 | XP_001071401.1 | 2 | 727 | ||
| XM_006243870.2 | XP_006243932.1 | 2 | 1873 | |||
| XM_008766597.1 | XP_008764819.1 | 3 | 453 | |||
| GenBank mRNA | FQ225859.1 | - | 2 | 300 | ||
| FQ232048.1 | - | 2 | 300 | |||

Lsmem2
Genomic Map