siRNA Details
| Gene Symbol: Lsmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001071401.3 | XP_001071401.1 | 2, 3 | 753 | ||
| XM_006243870.2 | XP_006243932.1 | 2, 3 | 1899 | |||
| XM_008766597.1 | XP_008764819.1 | 3, 4 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsmem2| Gene Symbol: Lsmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001071401.3 | XP_001071401.1 | 2, 3 | 753 | ||
| XM_006243870.2 | XP_006243932.1 | 2, 3 | 1899 | |||
| XM_008766597.1 | XP_008764819.1 | 3, 4 | 479 | |||