siRNA Details
| Gene Symbol: Mroh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729836.3 | XP_002729882.1 | 14 | 3053 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mroh6| Gene Symbol: Mroh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729836.3 | XP_002729882.1 | 14 | 3053 | ||