siRNA Details
| Gene Symbol: Arhgap35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271132.1 | NP_001258061.1 | 2 | 3669 | ||
| XM_006228412.2 | XP_006228474.1 | 2 | 4234 | |||
| XM_008758918.1 | XP_008757140.1 | 3 | 3494 | |||
| XM_008758919.1 | XP_008757141.1 | 2 | 3287 | |||

Arhgap35
Genomic Map