siRNA Details
| Gene Symbol: Ccdc166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130509.2 | NP_001123981.2 | 1 | 401 | ||
| NM_001163519.1 | NP_001156991.1 | 1 | 401 | |||
| GenBank mRNA | BC160829.1 | - | 1 | 401 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc166| Gene Symbol: Ccdc166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130509.2 | NP_001123981.2 | 1 | 401 | ||
| NM_001163519.1 | NP_001156991.1 | 1 | 401 | |||
| GenBank mRNA | BC160829.1 | - | 1 | 401 | ||