siRNA Details
| Gene Symbol: LOC100910223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764329.1 | XP_008762551.1 | 4 | 1471 | ||
| Gene Symbol: LOC691254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239357.2 | XP_006239419.1 | 5 | 522 | ||
| XM_006239358.1 | XP_006239420.1 | 5 | 419 | |||
| XM_006239359.2 | XP_006239421.1 | 4 | 429 | |||
| XM_006239361.2 | XP_006239423.1 | 3 | 320 | |||
| XM_008764309.1 | XP_008762531.1 | 4 | 2151 | |||
| XM_008764310.1 | XP_008762532.1 | 3 | 218 | |||
| XM_008764311.1 | XP_008762533.1 | 3 | 1927 | |||

LOC100910223
Genomic Map