siRNA Details
| Gene Symbol: Tmprss11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770103.1 | XP_008768325.1 | 6 | 477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11a| Gene Symbol: Tmprss11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770103.1 | XP_008768325.1 | 6 | 477 | ||