siRNA Details
| Gene Symbol: Iqsec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236890.2 | XP_006236952.1 | 3 | 1904 | ||
| XM_008763140.1 | XP_008761362.1 | 3 | 1904 | |||
| XM_008763141.1 | XP_008761363.1 | 3 | 1904 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqsec1| Gene Symbol: Iqsec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236890.2 | XP_006236952.1 | 3 | 1904 | ||
| XM_008763140.1 | XP_008761362.1 | 3 | 1904 | |||
| XM_008763141.1 | XP_008761363.1 | 3 | 1904 | |||