siRNA Details
| Gene Symbol: Tbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751162.3 | XP_003751210.1 | 6 | 942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbl2| Gene Symbol: Tbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751162.3 | XP_003751210.1 | 6 | 942 | ||