siRNA Details
| Gene Symbol: Fam60a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134711.1 | NP_001128183.1 | 3 | 437 | ||
| NM_001134712.1 | NP_001128184.1 | 3 | 273 | |||
| GenBank mRNA | BC168169.1 | - | 3 | 437 | 9024666 | |
| EV762285.1 | - | 3 | 304 | |||

Fam60a
Genomic Map