siRNA Details
| Gene Symbol: LOC100909526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765141.1 | XP_008763363.1 | 1, 2 | 1158 | ||
| XM_008765142.1 | XP_008763364.1 | 1, 2 | 1158 | |||
| XM_008765144.1 | XP_008763366.1 | 1, 2 | 1158 | |||
| Gene Symbol: Zfp709l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163063.1 | NP_001156535.1 | 2, 3 | 207 | ||
| XM_006240830.2 | XP_006240892.1 | 2, 3 | 227 | |||
| XM_008765131.1 | XP_008763353.1 | 2, 3 | 226 | |||

LOC100909526
Genomic Map