siRNA Details
| Gene Symbol: Il3ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248989.2 | XP_006249051.1 | 4 | 2764 | ||
| GenBank mRNA | BP502982.1 | - | 1 | 40 | ||
| FM042837.1 | - | 4 | 471 | |||
| FM046911.1 | - | 3 | 245 | |||
| FQ214452.1 | - | 1 | 67 | |||

Il3ra
Genomic Map