siRNA Details
| Gene Symbol: Znfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235641.1 | XP_006235703.1 | 14 | 6680 | ||
| XM_006235642.2 | XP_006235704.1 | 14 | 6420 | |||
| XM_006235643.2 | XP_006235705.1 | 14 | 6328 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znfx1| Gene Symbol: Znfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235641.1 | XP_006235703.1 | 14 | 6680 | ||
| XM_006235642.2 | XP_006235704.1 | 14 | 6420 | |||
| XM_006235643.2 | XP_006235705.1 | 14 | 6328 | |||