siRNA Details
| Gene Symbol: LOC686748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242580.2 | XP_006242642.2 | 1 | 406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC686748| Gene Symbol: LOC686748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242580.2 | XP_006242642.2 | 1 | 406 | ||