siRNA Details
| Gene Symbol: Mark4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191071.1 | NP_001178000.1 | 15, 16 | 1909 | ||
| XM_008758981.1 | XP_008757203.1 | 15, 16 | 1949 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mark4| Gene Symbol: Mark4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191071.1 | NP_001178000.1 | 15, 16 | 1909 | ||
| XM_008758981.1 | XP_008757203.1 | 15, 16 | 1949 | |||