siRNA Details
| Gene Symbol: Nkpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057808.5 | XP_001057808.2 | 5 | 3853 | ||
| XM_006228500.2 | XP_006228562.1 | 4 | 3406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkpd1| Gene Symbol: Nkpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001057808.5 | XP_001057808.2 | 5 | 3853 | ||
| XM_006228500.2 | XP_006228562.1 | 4 | 3406 | |||