siRNA Details
| Gene Symbol: BBS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252678.1 | NP_001239607.1 | 6 | 430 | ||
| NM_001320665.1 | NP_001307594.1 | 7 | 477 | |||
| NM_033028.4 | NP_149017.2 | 7 | 477 | |||
| NR_045565.1 | 8 | 535 | ||||
| NR_045566.1 | 7 | 790 | ||||
| XM_011521848.1 | XP_011520150.1 | 7 | 452 | |||
| XM_011521849.1 | XP_011520151.1 | 6 | 340 | |||
| XM_011521851.1 | XP_011520153.1 | 7 | 452 | |||
| XM_017022450.1 | XP_016877939.1 | 7 | 758 | |||
| XM_017022451.1 | XP_016877940.1 | 6 | 418 | |||
| XM_017022452.1 | XP_016877941.1 | 6 | 648 | |||
| XM_017022453.1 | XP_016877942.1 | 6 | 335 | |||
| XM_017022454.1 | XP_016877943.1 | 5 | 285 | |||
| GenBank mRNA | AF359281.1 | - | 7 | 449 | ||
| AK075321.1 | - | 7 | 441 | |||
| AK298922.1 | - | 8 | 507 | |||
| AK302224.1 | - | 7 | 744 | |||
| AK303706.1 | - | 6 | 425 | |||
| AK314507.1 | - | 7 | 449 | |||
| AY457143.1 | - | 7 | 549 | |||
| BC008923.2 | - | 4 | 254 | |||
| BC027624.2 | - | 7 | 447 | 4824276 | ||
| BE727117.1 | - | 6 | 418 | |||
| BI562463.1 | - | 7 | 472 | |||
| BX647855.1 | - | 6 | 401 | |||
| DB269838.1 | - | 7 | 470 | |||
| DC320069.1 | - | 7 | 472 | |||
| DC372967.1 | - | 7 | 477 | |||
| KU177936.1 | - | 7 | 428 | |||

BBS4
Genomic Map