siRNA Details
| Gene Symbol: Tex33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044289.1 | NP_001037754.1 | 2 | 600 | ||
| GenBank mRNA | BC079191.1 | - | 2 | 600 | 7131327 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex33| Gene Symbol: Tex33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044289.1 | NP_001037754.1 | 2 | 600 | ||
| GenBank mRNA | BC079191.1 | - | 2 | 600 | 7131327 | |