siRNA Details
| Gene Symbol: Bcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109422.1 | NP_001102892.1 | 1, 2 | 271 | ||
| XM_006228473.2 | XP_006228535.1 | 1, 2 | 364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl3| Gene Symbol: Bcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109422.1 | NP_001102892.1 | 1, 2 | 271 | ||
| XM_006228473.2 | XP_006228535.1 | 1, 2 | 364 | |||