siRNA Details
| Gene Symbol: Mbnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111064.1 | NP_001104534.1 | 3 | 916 | ||
| GenBank mRNA | CB757029.1 | - | 2 | 416 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mbnl2| Gene Symbol: Mbnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111064.1 | NP_001104534.1 | 3 | 916 | ||
| GenBank mRNA | CB757029.1 | - | 2 | 416 | ||