siRNA Details
| Gene Symbol: Gstt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109675.1 | NP_001103145.1 | 2 | 287 | ||
| GenBank mRNA | BC097423.1 | - | 2 | 271 | 7455039 | |
| Gene Symbol: LOC103694882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773850.1 | XP_008772072.1 | 2 | 310 | ||

Gstt4
Genomic Map