siRNA Details
Gene Symbol: LOC689713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047978.1 | NP_001041443.1 | 9 | 747 | ||
GenBank mRNA | AY539926.1 | - | 9 | 747 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC689713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047978.1 | NP_001041443.1 | 9 | 747 | ||
GenBank mRNA | AY539926.1 | - | 9 | 747 |