siRNA Details
Gene Symbol: Gxylt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001066466.4 | XP_001066466.1 | 5 | 1032 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gxylt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001066466.4 | XP_001066466.1 | 5 | 1032 |