siRNA Details
Gene Symbol: LOC690764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766033.1 | XP_008764255.1 | 545 |
Gene Symbol: LOC100911583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750446.2 | XP_003750494.2 | 2 | 629 |
