siRNA Details
| Gene Symbol: Ubap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243297.1 | XP_006243359.1 | 4 | 1406 | ||
| XM_006243298.1 | XP_006243360.1 | 5 | 1481 | |||
| XM_006243299.2 | XP_006243361.1 | 4 | 1287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubap1l| Gene Symbol: Ubap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243297.1 | XP_006243359.1 | 4 | 1406 | ||
| XM_006243298.1 | XP_006243360.1 | 5 | 1481 | |||
| XM_006243299.2 | XP_006243361.1 | 4 | 1287 | |||