siRNA Details
| Gene Symbol: Gpr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079710.1 | NP_001073178.1 | 2 | 1084 | ||
| GenBank mRNA | BC128782.1 | - | 2 | 1084 | 7375738 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr18| Gene Symbol: Gpr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079710.1 | NP_001073178.1 | 2 | 1084 | ||
| GenBank mRNA | BC128782.1 | - | 2 | 1084 | 7375738 | |