siRNA Details
| Gene Symbol: LOC686967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749071.3 | XP_003749119.2 | 4 | 756 | ||
| XM_008760279.1 | XP_008758501.1 | 3 | 903 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC686967| Gene Symbol: LOC686967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749071.3 | XP_003749119.2 | 4 | 756 | ||
| XM_008760279.1 | XP_008758501.1 | 3 | 903 | |||