siRNA Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031764.3 | NP_113952.2 | 7, 8 | 1112 | ||
| XM_006250226.2 | XP_006250288.1 | 8, 9 | 4409 | |||
| XM_006250227.2 | XP_006250289.1 | 10, 11 | 1744 | |||
| XM_006250228.2 | XP_006250290.1 | 9, 10 | 1525 | |||
| XM_006250229.2 | XP_006250291.1 | 9, 10 | 1473 | |||
| XM_008769761.1 | XP_008767983.1 | 9, 10 | 1487 | |||
| XM_008769762.1 | XP_008767984.1 | 9, 10 | 1419 | |||
| XM_008769763.1 | XP_008767985.1 | 9, 10 | 1725 | |||
| GenBank mRNA | BC161961.1 | - | 7, 8 | 1112 | 9027893 | |
| EV770183.1 | - | 4, 5 | 618 | |||

Ddr2
Genomic Map