siRNA Details
| Gene Symbol: LOC690821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766034.1 | XP_008764256.1 | 1 | 13 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690821| Gene Symbol: LOC690821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766034.1 | XP_008764256.1 | 1 | 13 | ||