siRNA Details
| Gene Symbol: NUP58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008564.1 | NP_001008564.1 | 5 | 862 | ||
| NM_014089.3 | NP_054808.1 | 6 | 898 | |||
| XM_005266600.3 | XP_005266657.1 | 6 | 1242 | |||
| XM_011535328.1 | XP_011533630.1 | 6 | 1242 | |||
| XM_011535329.2 | XP_011533631.1 | 7 | 1328 | |||
| XM_017020880.1 | XP_016876369.1 | 8 | 1410 | |||
| XM_017020881.1 | XP_016876370.1 | 7 | 1328 | |||
| GenBank mRNA | AB007870.1 | - | 6 | 795 | ||
| AK123955.1 | - | 7 | 732 | |||
| AK302946.1 | - | 5 | 757 | |||
| AK303409.1 | - | 5 | 666 | |||
| BC001104.2 | - | 6 | 770 | 3504343 | ||

NUP58
Genomic Map