siRNA Details
| Gene Symbol: Pcca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019330.1 | NP_062203.1 | 7 | 639 | ||
| XM_008770973.1 | XP_008769195.1 | 7 | 576 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcca| Gene Symbol: Pcca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019330.1 | NP_062203.1 | 7 | 639 | ||
| XM_008770973.1 | XP_008769195.1 | 7 | 576 | |||