siRNA Details
| Gene Symbol: Atp5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240845.2 | XP_006240907.1 | 2 | 308 | ||
| GenBank mRNA | BC161836.1 | - | 2 | 318 | 9098303 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp5d| Gene Symbol: Atp5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240845.2 | XP_006240907.1 | 2 | 308 | ||
| GenBank mRNA | BC161836.1 | - | 2 | 318 | 9098303 | |